Learn More
Merck Charged Durapore™ Membrane Filter, 0.22 μm
Recommended when maximum recovery is important
Brand: Merck CCGL09025
4731.44 NOK valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Additional Details : Weight : 0.01000kg
Description
Durapore™ membranes provide high flow rates and throughput, low extractables and broad chemical compatibility. Hydrophlic Durapore™ membrane binds far less protein than nylon, nitrocellulose, or PTFE membranes.Features & Benefits
- Available in several pore sizes (both hydrophilic and hydrophobic varieties) to suit your application needs
- Durapore™ membrane filters have very low protein binding to minimize interaction with your sample and maximize recovery
Specifications
Screen Filter | |
PVDF | |
90 mm |
0.22 μm | |
25Pack |