Learn More
Lenz Laborglasinstrumente™ Weighing Bottles
For the precise and secure weighing of samples in the laboratory. Lenz Laborglasinstrumente™ Weighing Bottles are manufactured from durable DURAN™ borosilicate glass.
Brand:  Lenz Laborglasinstrumente™ 05560504
Additional Details : Weight : 0.03900kg
 View more versions of this product
Product Code. 10008760
  • 235.00 NOK / Each
Estimated Shipment
14-02-2020
Add to basket
Description
Description
  • With interchangeable NS stopper or cap
  • With inscription label
  • With fused-in size (mm) and capacity (mL)
  • Available in high- or low-forms
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
Low
6Pack
Weighing Bottle
25 mm
Interchangeable NS stopper, inscription label
NS 40/12
Bottle