Learn More
DWK Life Sciences DURAN™ Jar, with shoulder and lid
5966.00 NOK valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Use promo code "23258" to get your promotional price.
Click to view available options
Capacity (Metric):
70 mL
250 mL
500 mL
Description
Ideally, containers that can be closed are used to store substances and materials. In this way, substances and materials can be protected from unwanted effects. These containers are available in various shapes such as cans, cylinders or jars. The choice of the vessel depends on the application and/or nature of the material.
- With shoulder and lid for safe storage and transport
- Fired-on, highly durable white ceramic
Specifications
Diameter (Metric) | 103 mm |
Capacity (Metric) | 250 mL |
For Use With (Application) | Storage and transport |
Material | Borosilicate Glass (3.3) |
Quantity | 10/Pk. |
Autoclavable | Autoclavable |