Learn More
DWK Life Sciences DURAN™ Exhibition glass
12492.50 NOK valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Use promo code "23258" to get your promotional price.
Click to view available options
Capacity (Metric):
100 mL
300 mL
600 mL
750 mL
1200 mL
Description
The multitude of different work steps in the laboratory environment demands a multitude of universally applicable glassware. These glass devices usually have a very simple shape and are characterized by their versatility. Exhibition glasses and museum jars are used to store materials and are often used in exhibitions. Here the objects on display can be preserved and then viewed without touching them.
- Overhead position of the specimen jar
- Tighter closure due to precise grinding
- Fired-on, highly durable white ceramic
Specifications
Diameter (Metric) | 90 mm |
Capacity (Metric) | 750 mL |
For Use With (Application) | Exhibition; Storage and transport of solid materials |
Material | Borosilicate Glass (3.3) |
Quantity | 10/Pk. |
Autoclavable | Autoclavable |