missing translation for 'onlineSavingsMsg'
Learn More

Doxycycline hyclate, USP grade, MP Biomedicals™

Catalog Number p-4405885
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
24390-14-5
C26H41ClN2O12
609.066
ISZOFWGVNMFENH-YDLUHMIOSA-N
131676157
(4S,4aR,5S,5aR,6R,12aR)-4-(dimethylamino)-1,5,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;ethanol;dihydrate;hydrochloride
CCO.CCO.CC1C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O.O.O.Cl
This item is not returnable. View return policy
24390-14-5
C26H41ClN2O12
609.066
ISZOFWGVNMFENH-YDLUHMIOSA-N
131676157
(4S,4aR,5S,5aR,6R,12aR)-4-(dimethylamino)-1,5,10,11,12a-pentahydroxy-6-methyl-3,12-dioxo-4a,5,5a,6-tetrahydro-4H-tetracene-2-carboxamide;ethanol;dihydrate;hydrochloride
CCO.CCO.CC1C2C(C3C(C(=O)C(=C(C3(C(=O)C2=C(C4=C1C=CC=C4O)O)O)O)C(=O)N)N(C)C)O.O.O.Cl
Type Doxycycline Hyclate
Physical Form Powder
Formula Weight 1025.89
Grade USP
Color Yellow
Quantity 1 g
Chemical Name or Material Doxycycline hyclate
compliance-icons
Product Identifier
  • Doxycycline Hyclate, USP
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : Doxycycline Hyclate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'