Learn More
Dowex™ 1X8 100-200 (Cl), Thermo Scientific Chemicals
Catalog Number p-7046418
531.62 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
50 g
250 g
1000 g
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
12627-85-9
C29H34ClN
432.05
MFCD00132718
BBQMUEOYPPPODD-UHFFFAOYSA-M
dowex 1x2 50-100 cl, dowex 1x8 50-100 cl
16212807
1,4-diethenylbenzene 4-ethenyl-N,N,N-trimethylanilinium ethenylbenzene chloride
*
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
12627-85-9
C29H34ClN
432.05
MFCD00132718
BBQMUEOYPPPODD-UHFFFAOYSA-M
dowex 1x2 50-100 cl, dowex 1x8 50-100 cl
16212807
1,4-diethenylbenzene 4-ethenyl-N,N,N-trimethylanilinium ethenylbenzene chloride
*
Dowex™ 1X8 100-200 (Cl) is used as an absorbent in solid phase extraction. It is used in anion-exchange chromatography for separation. It also finds application in fine chemical processing.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Dowex|r 1X8 100-200 (Cl) is used as an absorbent in solid phase extraction. It is used in anion-exchange chromatography for separation. It also finds application in fine chemical processing.
Solubility
Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Odor Amine-like
Quantity 50 g
Solubility Information Insoluble in water.
Chemical Name or Material Dowex™ 1X8 100-200 (Cl)