missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNAJC11 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
1611.84 NOK - 4593.76 NOK
Specifications
Antigen | DNAJC11 |
---|---|
Concentration | 0.14 mg/mL |
Applications | Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16887204
|
Proteintech
17331-1-AP-20UL |
20 μL |
1611.84 NOK
20µL |
Estimated Shipment: 06-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16877204
|
Proteintech
17331-1-AP-150UL |
150 μL |
4593.76 NOK
150µL |
Estimated Shipment: 06-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Belongs to the DNAJC11 family. Three named isoforms produced by alternative splicing.Specifications
DNAJC11 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q5U458, Q9NVH1 | |
230935, 362666, 55735 | |
DNAJC11 Fusion Protein Ag11310 | |
Primary | |
-20°C | |
DNAJC11 |
0.14 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
DNAJC11 | |
Dnajc11 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |