missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ DNAJA1 (Human) Recombinant Protein
Human DNAJA1 full-length ORF ( AAH08182, 1 a.a. - 397 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00003301-P01.25ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MVKETTYYDVLGVKPNATQEELKKAYRKLALKYHPDKNPNEGEKFKQISQAYEVLSDAKKRELYDKGGEQAIKEGGAGGGFGSPMDIFDMFFGGGGRMQRERRGKNVVHQLSVTLEDLYNGATRKLALQKNVICDKCEGRGGKKGAVECCPNCRGTGMQIRIHQIGPGMVQQIQSVCMECQGHGERISPKDRCKSCNGRKIVREKKILEVHIDKGMKDGQKITFHGEGDQEPGLEPGDIIIVLDQKDHAVFTRRGEDLFMCMDIQLVEALCGFQKPISTLDNRTIVITSHPGQIVKHGDIKCVLNEGMPIYRRPYEKGRLIIEFKVNFPENGFLSPDKLSLLEKLLPERKEVEETDEMDQVELVDFDPNQERRRHYNGEAYEDDEHHPRGGVQCQTS
Specifications
AAH08182 | |
DnaJ (Hsp40) homolog, subfamily A, member 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DNAJA1 | |
GST |
3301 | |
Wheat germ expression system | |
25 μg | |
DJ-2, DjA1, HDJ2, HSDJ, HSJ2, HSPF4, hDJ-2 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |