missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DLEC/CLEC4C/BDCA-2 Antibody (BIIB059) [Janelia Fluor™ 585], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28605JF585
This item is not returnable.
View return policy
Description
DLEC/CLEC4C/BDCA-2 Monoclonal antibody specifically detects DLEC/CLEC4C/BDCA-2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| DLEC/CLEC4C/BDCA-2 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| BDCA2, BDCA-2, BDCA2MGC125791, Blood dendritic cell antigen 2, blood dendritic cell antigen 2 protein, CD303, CD303 antigen, CLECSF11, CLECSF11PRO34150, CLECSF7, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 11, C-type (calcium dependent, carbohydrate-recognition domain) lectin, superfamilymember 7, C-type lectin domain family 4 member C, C-type lectin superfamily member 7, dendritic cell lectin b, Dendritic lectin, DLEC, DLECMGC125792, HECL, HECLMGC125793, MGC125789, PRO34150 | |
| CLEC4C | |
| 0.1 mL | |
| Dendritic Cell Markers | |
| 170482 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| BIIB059 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction