Learn More
Thermo Scientific Chemicals DL-Tryptophan, 99%
Catalog Number p-7044339
Click to view available options
Quantity:
5 g
25 g
100 g
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
54-12-6
C11H12N2O2
204.23
MFCD00064339
QIVBCDIJIAJPQS-UHFFFAOYSA-N
dl-tryptophan, 2-amino-3-1h-indol-3-yl propanoic acid, racemic tryptophan, dl-trytophane, dl-trytophan, +--tryptophan, h-dl-trp-oh, dl-3beta-indolylalanine, dl-tryptophane, tryptophan .
1148
CHEBI:27897
2-amino-3-(1H-indol-3-yl)propanoic acid
NC(CC1=CNC2=CC=CC=C12)C(O)=O
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
54-12-6
C11H12N2O2
204.23
MFCD00064339
QIVBCDIJIAJPQS-UHFFFAOYSA-N
dl-tryptophan, 2-amino-3-1h-indol-3-yl propanoic acid, racemic tryptophan, dl-trytophane, dl-trytophan, +--tryptophan, h-dl-trp-oh, dl-3beta-indolylalanine, dl-tryptophane, tryptophan .
1148
CHEBI:27897
2-amino-3-(1H-indol-3-yl)propanoic acid
NC(CC1=CNC2=CC=CC=C12)C(O)=O
DL-Tryptophan, CAS # 54-12-6, is an α-amino acid that is commonly used in the synthesis of proteins. It is an essential amino acid precursor for the formation of serotonin, melatonin, and vitamin B3.
DL-Tryptophan is an amino acid precursor of serotonin and melatonin. It is a dietary supplement for use as an antidepressant, anxiolytic, and sleep aid. DL-tryptophan also can be used as feed nutrition enhancer, antioxidants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
General Description
• DL-Tryptophan is an α-amino acid, and an essential amino acid precursor of serotonin, melatonin, and vitamin B3
Application
• DL-Tryptophan shows antioxidant activity• It has a role as a Daphnia magna metabolite
• It has been explored in in vitro studies to evaluate the antimicrobial and immunoregulatory functions on indoleamine 2,3-dioxygenase
• It has been used as a medium supplement in Psilocybe atrobrunnea and Psilocybe serbica for the synthesis of compounds
• It can be used as a medium supplement for culturing yeast
TRUSTED_SUSTAINABILITY
Melting Point ∽280°C (decomposition)
Beilstein 86196
Solubility Information Soluble I water. 10g/L (20°C),Hcl and alkalies. Sparingly soluble in alcohol.
Formula Weight 204.23
Percent Purity 99%
Quantity 5 g
Chemical Name or Material DL-Tryptophan