missing translation for 'onlineSavingsMsg'
Learn More

DL-Pantolactone, 97%, Thermo Scientific Chemicals

Catalog Number p-3771069
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
79-50-5
C6H10O3
130.14
MFCD00064333
SERHXTVXHNVDKA-UHFFFAOYSA-N
pantolactone, dl-pantolactone, 3-hydroxy-4,4-dimethyldihydrofuran-2 3h-one, pantoyl lactone, pantoic lactone, dl-pantoyl lactone, d---pantolactone, 2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl, dihydro-3-hydroxy-4,4-dimethyl-2 3h-furanone, pantolyl lactone
989
3-hydroxy-4,4-dimethyloxolan-2-one
CC1(COC(=O)C1O)C
This item is not returnable. View return policy
79-50-5
C6H10O3
130.14
MFCD00064333
SERHXTVXHNVDKA-UHFFFAOYSA-N
pantolactone, dl-pantolactone, 3-hydroxy-4,4-dimethyldihydrofuran-2 3h-one, pantoyl lactone, pantoic lactone, dl-pantoyl lactone, d---pantolactone, 2 3h-furanone, dihydro-3-hydroxy-4,4-dimethyl, dihydro-3-hydroxy-4,4-dimethyl-2 3h-furanone, pantolyl lactone
989
3-hydroxy-4,4-dimethyloxolan-2-one
CC1(COC(=O)C1O)C

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 74.0°C to 78.0°C
Boiling Point 246.0°C
Flash Point 122°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: soluble.
Formula Weight 130.14
Percent Purity 97%
Chemical Name or Material DL-Pantolactone
compliance-icons
Product Identifier
  • DL-Pantolactone
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'