Learn More
DL-Lactide, 99%, Thermo Scientific Chemicals
350.41 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
50 g
Description
DL-Lactide is used to produce 2-hydroxy-propionic acid 1-(1-phenyl-ethoxycarbonyl)-ethyl ester. It acts as an important raw material and an intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyes. It is involved in enzymatic alcoholysis to produce both alkyl (R)-lactates and alkyl (S,S)-lactyllactates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
DL-Lactide is used to produce 2-hydroxy-propionic acid 1-(1-phenyl-ethoxycarbonyl)-ethyl ester. It acts as an important raw material and an intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyes. It is involved in enzymatic alcoholysis to produce both alkyl (R)-lactates and alkyl (S,S)-lactyllactates.
Solubility
Soluble in chloroform and methanol. Slightly soluble in benzene.
Notes
Moisture senstive. Store in a cool place. Incompatible with acids, bases and strong oxidizing agents.
DL-Lactide is used to produce 2-hydroxy-propionic acid 1-(1-phenyl-ethoxycarbonyl)-ethyl ester. It acts as an important raw material and an intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyes. It is involved in enzymatic alcoholysis to produce both alkyl (R)-lactates and alkyl (S,S)-lactyllactates.
Solubility
Soluble in chloroform and methanol. Slightly soluble in benzene.
Notes
Moisture senstive. Store in a cool place. Incompatible with acids, bases and strong oxidizing agents.
Specifications
Melting Point | 122°C to 126°C |
Boiling Point | 138°C to 142°C (8 mmHg) |
Quantity | 10 g |
Beilstein | 82121 |
Solubility Information | Soluble in chloroform and methanol. Slightly soluble in benzene. |
Formula Weight | 144.13 |
Percent Purity | 99% |
Chemical Name or Material | DL-Lactide |