Learn More
DL-3-Amino-3-phenylpropionic acid, 97%, ACROS Organics™
Product Code. 10720871 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
Unit Size:
25g
Product Code. 10720871
Brand: Thermo Scientific Acros 170380250
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
CAS 614-19-7
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C9H11NO2
Packaging Glass Bottle
Linear Formula C6H5CH(NH2)CH2CO2H
MDL Number MFCD00008064,MFCD01076238
Beilstein 14, 493
Synonym 3-amino-3-phenylpropionic acid, dl-3-amino-3-phenylpropionic acid, dl-beta-phenylalanine, beta-phenyl-beta-alanine, 3-phenyl-dl-beta-alanine, h-dl-beta-phe-oh, benzenepropanoic acid, beta-amino, 3-amino-3-phenyl-propionic acid, 3a3ppa, beta-aminohydrocinnamic acid
InChI Key UJOYFRCOTPUKAK-UHFFFAOYNA-N
SMILES NC(CC(O)=O)C1=CC=CC=C1
IUPAC Name 3-amino-3-phenylpropanoic acid
Molecular Weight (g/mol) 165.19
Quantity 25 g
PubChem CID 69189
ChEBI CHEBI:68528
Formula Weight 165.19
Percent Purity ≥96.0%
Grade Technical
Physical Form Powder
Chemical Name or Material DL-3-Amino-3-phenylpropionic acid, tech., 90%