missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Divinyl sulfone, 97%, stab. with 0.05% hydroquinone, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
Divinyl sulfone acts as a cross-linking reagent for agarose gels. It is a monomer, which is used in the production of polymers with diols, urea and malonic esters. Further, it serves as a shrinkage control agent in textiles. In addition to this, it is used in the preparation of large class of fiber-reactive dyestuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Divinyl sulfone acts as a cross-linking reagent for agarose gels. It is a monomer, which is used in the production of polymers with diols, urea and malonic esters. Further, it serves as a shrinkage control agent in textiles. In addition to this, it is used in the preparation of large class of fiber-reactive dyestuffs.
Solubility
Miscible with water.
Notes
Incompatible with strong oxidizing agents, strong acids, reducing agents, alkali metals and strong bases.
Divinyl sulfone acts as a cross-linking reagent for agarose gels. It is a monomer, which is used in the production of polymers with diols, urea and malonic esters. Further, it serves as a shrinkage control agent in textiles. In addition to this, it is used in the preparation of large class of fiber-reactive dyestuffs.
Solubility
Miscible with water.
Notes
Incompatible with strong oxidizing agents, strong acids, reducing agents, alkali metals and strong bases.
Specifications
Melting Point | ∼-26°C |
Density | 1.177 |
Boiling Point | 233°C to 234°C |
Flash Point | 102°C (215°F) |
Refractive Index | 1.476 |
Quantity | 10 g |
UN Number | UN2927 |
Beilstein | 1071329 |
Solubility Information | Miscible with water. |
Formula Weight | 118.16 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'