missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diphenyl selenide, 97%, Thermo Scientific Chemicals
320.41 NOK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
1 g
5 g
25 g
Description
Diphenyl selenide is used for anti-selective Michael addition of thiols and their analogs to nitro-olefins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Diphenyl selenide is used for anti-selective Michael addition of thiols and their analogs to nitro-olefins.
Solubility
Immiscible with water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.
Diphenyl selenide is used for anti-selective Michael addition of thiols and their analogs to nitro-olefins.
Solubility
Immiscible with water.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place.
Specifications
Melting Point | 3°C |
Density | 1.351 |
Boiling Point | 165°C to 167°C (12 mmHg) |
Odor | Unpleasant |
Quantity | 1 g |
UN Number | UN3440 |
Solubility Information | Immiscible with water. |
Formula Weight | 233.17 |
Percent Purity | 98% |
Chemical Name or Material | Diphenyl selenide |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'