Learn More
Alfa Aesar™ Dimethyl octafluoroadipate, 94%
Brand: Alfa Aesar™ L16752.14
Additional Details : CAS Number : 3107-98-0 Weight : 0.02500kg
Quantity | 25g |
---|
Chemical Identifiers
3107-98-0 | |
318.119 | |
XPXVIIILXUOEQA-UHFFFAOYSA-N | |
2737117 | |
COC(=O)C(C(C(C(C(=O)OC)(F)F)(F)F)(F)F)(F)F |
C8H6F8O4 | |
MFCD00042267 | |
dimethyl octafluoroadipate, dimethyl perfluoroadipate, hexanedioic acid, octafluoro-, dimethyl ester, 1,6-dimethyl 2,2,3,3,4,4,5,5-octafluorohexanedioate, dimethyloctafluoroadipate, acmc-1cson, dimethyl octafluoroadipinate, dimethyl octafluorohexanedioate, perfluoroadipic acid dimethyl ester, octafluoroadipic acid dimethyl ester | |
dimethyl 2,2,3,3,4,4,5,5-octafluorohexanedioate |
Specifications
Dimethyl octafluoroadipate | |
1.3475 | |
25g | |
C8H6F8O4 | |
MFCD00042267 | |
dimethyl octafluoroadipate, dimethyl perfluoroadipate, hexanedioic acid, octafluoro-, dimethyl ester, 1,6-dimethyl 2,2,3,3,4,4,5,5-octafluorohexanedioate, dimethyloctafluoroadipate, acmc-1cson, dimethyl octafluoroadipinate, dimethyl octafluorohexanedioate, perfluoroadipic acid dimethyl ester, octafluoroadipic acid dimethyl ester | |
COC(=O)C(C(C(C(C(=O)OC)(F)F)(F)F)(F)F)(F)F | |
318.119 | |
318.12 |
1.547 | |
120°C (15mmHg) | |
3107-98-0 | |
CH3O2C(CF2)4CO2CH3 | |
1916112 | |
XPXVIIILXUOEQA-UHFFFAOYSA-N | |
dimethyl 2,2,3,3,4,4,5,5-octafluorohexanedioate | |
2737117 | |
94% |