Learn More
Dimethyl maleate, 96%, Thermo Scientific Chemicals
Description
Dimethyl maleate is acts as a dienophile involved in ultrasonic irradiation promoted Diels-Alder reaction with substituted furans. It is widely used as an intermediate and additive for pharmaceuticals, copolymers, pigments, plastics, paints, adhesives and agrochemicals. As an internal modifier, it increases the glass transition temperature of styrene or vinyl chloride polymer. It plays a vital role to improve the hardness and toughness of polymer films such as copolymers of vinyl acetate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Dimethyl maleate is acts as a dienophile involved in ultrasonic irradiation promoted Diels-Alder reaction with substituted furans. It is widely used as an intermediate and additive for pharmaceuticals, copolymers, pigments, plastics, paints, adhesives and agrochemicals. As an internal modifier, it increases the glass transition temperature of styrene or vinyl chloride polymer. It plays a vital role to improve the hardness and toughness of polymer films such as copolymers of vinyl acetate.
Solubility
Miscible with water.
Notes
Incompatible with acids, bases, oxidizing agents and reducing agents.
Specifications
Melting Point | -19°C |
Density | 1.151 |
Boiling Point | 204°C to 205°C |
Flash Point | 91°C (195°F) |
Refractive Index | 1.442 |
Quantity | 100 g |
UN Number | UN2810 |
Beilstein | 471705 |
Solubility Information | Miscible with water. |
Formula Weight | 144.13 |
Show More |
RUO – Research Use Only