Learn More
Diethyl benzylphosphonate, 99%, Thermo Scientific Chemicals
Catalog Number p-4258948
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
1080-32-6
C11H17O3P
228.23
AIPRAPZUGUTQKX-UHFFFAOYSA-N
diethyl benzylphosphonate, benzylphosphonic acid diethyl ester, diethoxyphosphonomethylbenzene, diethyl phosphonate, benzyl, phosphonic acid, phenylmethyl-, diethyl ester, unii-t6d4qwt656, diethylbenzylphosphonate, phosphonic acid, benzyl-, diethyl ester, diethyl phenylmethyl phosphonate, phosphonic acid, p-phenylmethyl-, diethyl ester
14122
diethoxyphosphorylmethylbenzene
CCOP(=O)(CC1=CC=CC=C1)OCC
1080-32-6
C11H17O3P
228.23
AIPRAPZUGUTQKX-UHFFFAOYSA-N
diethyl benzylphosphonate, benzylphosphonic acid diethyl ester, diethoxyphosphonomethylbenzene, diethyl phosphonate, benzyl, phosphonic acid, phenylmethyl-, diethyl ester, unii-t6d4qwt656, diethylbenzylphosphonate, phosphonic acid, benzyl-, diethyl ester, diethyl phenylmethyl phosphonate, phosphonic acid, p-phenylmethyl-, diethyl ester
14122
diethoxyphosphorylmethylbenzene
CCOP(=O)(CC1=CC=CC=C1)OCC
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For C-C Bond Formation (Olefination)
TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Density 1.0900g/mL
Boiling Point 106.0°C to 108.0°C (1.0 mmHg)
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula (C2H5O)2P(O)CH2C6H5
Refractive Index 1.4950 to 1.4980
Quantity 25 g
Specific Gravity 1.09
Formula Weight 228.23
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Diethyl benzylphosphonate
Show More Show Less
compliance-icons
Product Identifier
  • Diethyl benzylphosphonate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.