Learn More
BioTek Instruments Half-size Dichroic Mirror
For use with Cytation 5, Synergy H1 and Synergy LX
Brand:  BioTek Instruments 7138545
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12542787
  • 4091.00 NOK / Each
Estimated Shipment: 06-12-2021
Description
Description
For use with Cytation 5, Synergy H1 and Synergy LX
Specifications
Specifications
545 nm
Cytation 3, Cytation 5, Synergy 2, Synergy H1, Ex. 512-535, Em. 555-578
Dichroic Mirror