Learn More
Diaion™ SA12A(Cl), ion exchange resin, moderately cross-linked, strongly basic gel type I, 1.3 meq/ml on PS-DVB, Thermo Scientific™
Used in demineralization, decolorization, metal recovery, high purity water
Brand:  Thermo Scientific Alfa Aesar 046434.A1
Additional Details : Weight : 1.00000kg
Product Code. 15454045
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Description
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Specifications
300-1180μm Beads
Moderately cross-linked, strongly basic gel type I, 1.3meq/mL on PS-DVB
1 kg
Diaion™ SA12A (Cl), ion exchange resin
300 to 1180μm