Learn More
Di-tert-butyl phosphite, 96%, Thermo Scientific Chemicals
Catalog Number p-4813464
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
13086-84-5
C8H18O3P
193.20
MFCD00014999,MFCD00014999
GEBLOQXLELCEEO-UHFFFAOYSA-N
di-tert-butyl phosphite, di-tert-butylphosphite, phosphorous acid, bis 1,1-dimethylethyl ester
21959072
ditert-butyl phosphite
CC(C)(C)O[P+](=O)OC(C)(C)C
13086-84-5
C8H18O3P
193.20
MFCD00014999,MFCD00014999
GEBLOQXLELCEEO-UHFFFAOYSA-N
di-tert-butyl phosphite, di-tert-butylphosphite, phosphorous acid, bis 1,1-dimethylethyl ester
21959072
ditert-butyl phosphite
CC(C)(C)O[P+](=O)OC(C)(C)C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Density 0.9950g/mL
Boiling Point 66°C to 68°C (0.5 mmHg)
Flash Point 163°C
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula [(CH3)3CO]2P(O)H
Quantity 1 g
Specific Gravity 0.995
Solubility Information Solubility in water: immiscible
Formula Weight 193.2
Percent Purity 96%
Chemical Name or Material Di-tert-butyl phosphite
Show More Show Less
compliance-icons
Product Identifier
  • Di-tert-butyl phosphite
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.