Learn More
Di-n-propyl-d7-amine (mono-propyl-d7), CDN
Click to view available options
Quantity:
0.05 g
Unit Size:
0.05g
Specifications
Specifications
| Chemical Name or Material | Dipropylamine-1,1,2,2,3,3,3-D7 |
| CAS | 63220-61-1 |
| Molecular Formula | C6 D7 H8 N |
| Quantity | 0.05 g |
| Synonym | 1-Propan-1,1,2,2,3,3,3-d7-amine, N-propyl- (9 CI), N-Propyl-1-propan-1,1,2,2,3,3,3-d7-amine, Dipropylamine-d7, Dipropylamine-1,1,2,2,3,3,3-D7, Di-n-propyl-d7-amine (mono-propyl-d7) |
| InChI Formula | InChI=1 S/C6H15N/c1-3-5-7-6-4-2/h7H,3-6H2,1-2H3/i1D3,3D2,5D2 |
| SMILES | [2 H]C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])NCCC |
| IUPAC Name | 1,1,2,2,3,3,3-heptadeuterio-N-propylpropan-1-amine |
| Molecular Weight (g/mol) | 108.2331 |
| Formula Weight | 108.1644 g/mol |
| Show More |