Learn More
DEXI Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA559605
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: IVLNVGLVFL PEDMDQALVD LGVLSDPGSG LYDADSELDV FDAYLE Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 96%.
DEXI Polyclonal antibody specifically detects DEXI in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.Specifications
DEXI | |
Polyclonal | |
Unconjugated | |
Dexi | |
1810029J14Rik; AI836170; AW413143; D16Bwg0586e; Dexamethasone-induced protein; dexamethasone-induced transcript; Dexi; Dexi homolog; Myle; Protein MYLE; RGD1564938 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
28955 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O95424 | |
Dexi | |
Recombinant protein corresponding to Human DEXI | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only