Learn More
Desonide, MedChemExpress
Product Code. 16498186
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
Product Code. 16498186
Brand: Medchem Express HYB0248
MedChemExpress Desonide is a nonfluorinated corticosteroid anti-inflammatory agent used topically for dermatoses.
Desonide is a nonfluorinated corticosteroid anti-inflammatory agent used topically for dermatoses.
Specifications
Chemical Name or Material Desonide
CAS 638-94-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C24H32O6
Quantity 100 mg
Solubility Information DMSO : ≥ 100 mg/mL (240.09 mM)
SMILES C[C@@]12[C@@]3(C(CO)=O)[C@@](OC(C)(O3)C)([H])C[C@@]1([H])[C@]4([H])CCC5=CC(C=C[C@]5(C)[C@@]4([H])[C@@H](O)C2)=O
Molecular Weight (g/mol) 416.51
Formula Weight 416.51
Percent Purity 99.45%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.