missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Decorin Antibody (DCN/7031R) [Janelia Fluor™ 585], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14026JF585
This item is not returnable.
View return policy
Description
Decorin Monoclonal antibody specifically detects Decorin in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| Decorin | |
| Monoclonal | |
| Janelia Fluor 585 | |
| Bone proteoglycan II, CSCD, decorin, decorin proteoglycan, dermatan sulphate proteoglycans II, DSPG2, PG40, PGII, PG-S2, proteoglycan core protein, SLRR1BPGS2, small leucine-rich protein 1B | |
| Recombinant human Decorin protein fragment (around aa212-336) (exact sequence is proprietary) (Uniprot: P07585) | |
| 0.1 mL | |
| Cancer | |
| 1634 | |
| Store at 4°C in the dark. | |
| IgG κ |
| Immunohistochemistry (Paraffin) | |
| DCN/7031R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction