Learn More
Trajan Scientific and Medical™ DANI Liners
Provides better analysis for a wide range of compounds. Trajan Scientific and Medical DANI Liners has unique deactivation process for SGE standard inlets.
Brand: Trajan Scientific and Medical™ 092500
Additional Details : Weight : 0.01000kg
Description
- Each batch of liners tested for activity
- Manufactured from borosilicate glass
- Color coded by geometry for easy identification
- For use with DANI instruments
Specifications
Straight-through Liner with quartz wool | |
DANI Liners | |
70 mm | |
6 mm |
Deactivated Liners For DANI GCs | |
5/Pk. | |
4 mm | |
Straight Liner with Wool for DANI |