missing translation for 'onlineSavingsMsg'
Learn More

Uvex™ D500 Foam Cut Protection Gloves

Click to view available options
Size:
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Bamboo TwinFlex™ technology provides level D cut protection, while SoftGrip foam coating on the palm and fingertips enhances grip capabilities

  • Bamboo TwinFlex™ Technology provides level D cut protection
  • Highly abrasion resistant for extended glove life and cost efficiency
  • High-performance elastomer (HPE) and SoftGrip foam coating for enhanced grip
  • Increased tactile sensitivity for part handling & assembly
  • 3D Ergo Technology ensures a secure fit
  • Free of harmful substances in accordance with OEKO-TEX™ Standard 100
Certifications/Compliance OEKO-TEX™ Standard 100
Applications Industrial
Coating Material High Performance Elastomer (HPE), SoftGrip Foam
Color Green, Black
Material Dyneema™, Polyamide, Bamboo
Size 10
EN388 Standard 4X42D
For Use With (Application) Automotive, Construction, Glass Handling, Maintenance, and Touchscreens
Gender Unisex
Glove Cuff Style Knit
Series D500 Foam
Show More Show Less