missing translation for 'onlineSavingsMsg'
Learn More

D-myo-Inositol-4-phosphate ammonium salt, 98%, Thermo Scientific™

Catalog Number 15448599
Click to view available options
Quantity:
0.1 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
CAS 142760-33-6
Color White
Molecular Formula C6H19N2O9P
MDL Number MFCD00135425
Quantity 0.1 mg
Synonym Ins(4)P1 ammonium salt; 4-IP1 ammonium salt
InChI Key HULWABRHGYHABN-FGPSAQHISA-N
SMILES C1(C(C(C(C(C1O)O)OP(=O)(O)O)O)O)O.N.N
IUPAC Name azane;[(2S,3S,5S,6R)-2,3,4,5,6-pentahydroxycyclohexyl] dihydrogen phosphate
Molecular Weight (g/mol) 294.197
PubChem CID 131873651
Formula Weight 276.16
Percent Purity 98%
Physical Form Lyophilized Powder
Chemical Name or Material D-myo-Inositol-4-phosphate ammonium salt
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'