missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Maltose monohydrate, 95 to 98%, MP Biomedicals™

Brand:  MP Biomedicals 0210224190

Additional Details : CAS Number : 6363-53-7 Weight : 0.01000kg

Quantity 500g

 View more versions of this product

Product Code. 11389109

  • 4094.00 NOK / 500g


Estimated Shipment
23-02-2021
Add to basket

Description

Description

Molecular biology
Specifications

Chemical Identifiers

6363-53-7
C12H22O11.H2O
MFCD00149343
d-+-maltose monohydrate, unii-dm477ee40d, 2r,3r,4r,5r-2,3,5,6-tetrahydroxy-4-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy hexanal hydrate, beta-maltose monohydrate, d-+-maltosemonohydrate, 69-79-4 anhydrous, d +-maltose monohydrate, d-+-maltose monohydrate, puriss., d-+-maltose monohydrate, analytical standard, d-+-maltose monohydrate, bioxtra

Specifications

D-(+)-Maltose monohydrate
White
6363-53-7
C12H22O11.H2O
d-+-maltose monohydrate, unii-dm477ee40d, 2r,3r,4r,5r-2,3,5,6-tetrahydroxy-4-2r,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl oxy hexanal hydrate, beta-maltose monohydrate, d-+-maltosemonohydrate, 69-79-4 anhydrous, d +-maltose monohydrate, d-+-maltose monohydrate, puriss., d-+-maltose monohydrate, analytical standard, d-+-maltose monohydrate, bioxtra
Solid
1.54g/mL
500g
95 to 98%
MFCD00149343
360.32
Videos
SDS
Documents