Learn More
D-Gluconic acid, calcium salt, 99%, Thermo Scientific Chemicals
Catalog Number p-3734379
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Plastic drum
299-28-5
C12H22CaO14
430.372
MFCD00064209
NEEHYRZPVYRGPP-IYEMJOQQSA-L
calcium gluconate, calcium d-gluconate, calciofon, calglucon, glucobiogen, ebucin, calcicol, calcipur, calglucol, dragocal
9290
calcium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
C(C(C(C(C(C(=O)[O-])O)O)O)O)O.C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Ca+2]
299-28-5
C12H22CaO14
430.372
MFCD00064209
NEEHYRZPVYRGPP-IYEMJOQQSA-L
calcium gluconate, calcium d-gluconate, calciofon, calglucon, glucobiogen, ebucin, calcicol, calcipur, calglucol, dragocal
9290
calcium;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate
C(C(C(C(C(C(=O)[O-])O)O)O)O)O.C(C(C(C(C(C(=O)[O-])O)O)O)O)O.[Ca+2]
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Color White
Loss on Drying 2.0 % max. (1 g, 105°C, 16 hrs)
Infrared Spectrum Authentic
Assay Percent Range 0.99
Packaging Plastic bottle
Linear Formula (HOCH2[CH(OH)]4CO2)2Ca
Quantity 250 g
Beilstein 03, 544
Merck Index 15, 1671
Specific Rotation Condition + 10.20 (22.00°C c=1,H2O)
Specific Rotation + 10.20
Formula Weight 430.38
Percent Purity 0.99
Physical Form Crystalline Powder or Crystals
Chemical Name or Material D-Gluconic acid, calcium salt