Learn More
D-Aspartic acid 1-methyl ester, 98%, Thermo Scientific Chemicals
Catalog Number p-7055150
Click to view available options
Quantity:
1 g
5 g
25 g
65414-78-0
C5H9NO4
147.13
MFCD03424227
SWWBMHIMADRNIK-GSVOUGTGSA-N
h-d-asp-ome, r-3-amino-4-methoxy-4-oxobutanoic acid, d-asp-ome, d-aspartic acid 1-methyl ester, d-aspartic acid,1-methyl ester, 3r-3-amino-4-methoxy-4-oxobutanoic acid
7020023
(3R)-3-amino-4-methoxy-4-oxobutanoic acid
COC(=O)C(CC(=O)O)N
65414-78-0
C5H9NO4
147.13
MFCD03424227
SWWBMHIMADRNIK-GSVOUGTGSA-N
h-d-asp-ome, r-3-amino-4-methoxy-4-oxobutanoic acid, d-asp-ome, d-aspartic acid 1-methyl ester, d-aspartic acid,1-methyl ester, 3r-3-amino-4-methoxy-4-oxobutanoic acid
7020023
(3R)-3-amino-4-methoxy-4-oxobutanoic acid
COC(=O)C(CC(=O)O)N
D-Aspartic acid 1-methyl ester is used as a intermediate for pharmaceutical and chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
D-Aspartic acid 1-methyl ester is used as a intermediate for pharmaceutical and chemical research.
Notes
Refrigerate. Keep container tightly closed. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Formula Weight 147.13
Percent Purity 98%
Quantity 1 g
Physical Form Solid
Chemical Name or Material D-Aspartic acid 1-methyl ester