missing translation for 'onlineSavingsMsg'
Learn More

D-Asparagine monhydrate, 100.5%, MP Biomedicals™

Catalog Number p-4403848
Click to view available options
Quantity:
500 g
1 kg
This item is not returnable. View return policy
5794-24-1
C4H10N2O4
150.134
RBMGJIZCEWRQES-HSHFZTNMSA-N
d-asparagine monohydrate, d-asparagine hydrate, r-2,4-diamino-4-oxobutanoic acid hydrate, d-asparagine, monohydrate, d-asparaginemonohydrate, d---asparagine monohydrate, d-aspartic acid 4-amide, r---2-aminosuccinamic acid, d--asparagine monohydrate, r-2-aminosuccinic acid 4-amide
16211197
(2R)-2,4-diamino-4-oxobutanoic acid;hydrate
C(C(C(=O)O)N)C(=O)N.O
This item is not returnable. View return policy
5794-24-1
C4H10N2O4
150.134
RBMGJIZCEWRQES-HSHFZTNMSA-N
d-asparagine monohydrate, d-asparagine hydrate, r-2,4-diamino-4-oxobutanoic acid hydrate, d-asparagine, monohydrate, d-asparaginemonohydrate, d---asparagine monohydrate, d-aspartic acid 4-amide, r---2-aminosuccinamic acid, d--asparagine monohydrate, r-2-aminosuccinic acid 4-amide
16211197
(2R)-2,4-diamino-4-oxobutanoic acid;hydrate
C(C(C(=O)O)N)C(=O)N.O
Assay Percent Range 1.005
Formula Weight 150.1
Quantity 500 g
Physical Form Crystalline
Chemical Name or Material D-Asparagine monhydrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'