missing translation for 'onlineSavingsMsg'
Learn More

D-Alanine methyl ester hydrochloride, 98%, Thermo Scientific Chemicals

Catalog Number p-4228292
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
14316-06-4
C4H10ClNO2
139.58
MFCD00066141
IYUKFAFDFHZKPI-UHFFFAOYNA-N
d-alanine methyl ester hydrochloride, methyl d-alaninate hydrochloride, h-d-ala-ome.hcl, methyl d-alaninate hcl, methyl 2r-2-aminopropanoate hydrochloride, h-d-ala-ome hcl, r-methyl 2-aminopropanoate hydrochloride, d-alanine methylester hydrochloride, d-alanine, methyl ester, hydrochloride, pubchem10885
11182647
methyl (2R)-2-aminopropanoate;hydrochloride
[H+].[Cl-].COC(=O)C(C)N
This item is not returnable. View return policy
14316-06-4
C4H10ClNO2
139.58
MFCD00066141
IYUKFAFDFHZKPI-UHFFFAOYNA-N
d-alanine methyl ester hydrochloride, methyl d-alaninate hydrochloride, h-d-ala-ome.hcl, methyl d-alaninate hcl, methyl 2r-2-aminopropanoate hydrochloride, h-d-ala-ome hcl, r-methyl 2-aminopropanoate hydrochloride, d-alanine methylester hydrochloride, d-alanine, methyl ester, hydrochloride, pubchem10885
11182647
methyl (2R)-2-aminopropanoate;hydrochloride
[H+].[Cl-].COC(=O)C(C)N

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 60.4°C to 62.1°C
Color White
Elemental Analysis 33.56 to 35.28% C, 9.78 to 10.28% N
Infrared Spectrum Authentic
Assay Percent Range 97.5 to max. 102.5% min. (Argentometry)
Packaging Glass bottle
Quantity 5 g
Specific Rotation Condition - 12.50 (19.00°C +/-2.5,c=2,MeOH)
Specific Rotation -12.5
Formula Weight 139.58
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material D-Alanine methyl ester hydrochloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'