missing translation for 'onlineSavingsMsg'
Learn More

D-(-)-2-Phenylglycine chloride hydrochloride, 98%, Thermo Scientific™

Catalog Number p-7052866
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
39878-87-0
C8H9Cl2NO
206.066
MFCD00039094
GVVFCAFBYHYGEE-OGFXRTJISA-N
r-2-amino-2-phenylacetyl chloride hydrochloride, r---2-phenylglycine chloride hydrochloride, unii-07v2dd878h, 2r-2-amino-2-phenylacetyl chloride hydrochloride, --alpha-chloroformyl benzylammonium chloride, benzeneacetyl chloride, alpha-amino-, hydrochloride, alphar, d---phenylglycine acid chloride, hydrochloride, r-alpha-aminobenzeneacetyl chloride, hydrochloride, r---2-phenylglycine chloride hcl, 2r-amino phenyl acetoyl chloride hydrochloride 1:1
170253
(2R)-2-amino-2-phenylacetyl chloride;hydrochloride
C1=CC=C(C=C1)C(C(=O)Cl)N.Cl
This item is not returnable. View return policy
39878-87-0
C8H9Cl2NO
206.066
MFCD00039094
GVVFCAFBYHYGEE-OGFXRTJISA-N
r-2-amino-2-phenylacetyl chloride hydrochloride, r---2-phenylglycine chloride hydrochloride, unii-07v2dd878h, 2r-2-amino-2-phenylacetyl chloride hydrochloride, --alpha-chloroformyl benzylammonium chloride, benzeneacetyl chloride, alpha-amino-, hydrochloride, alphar, d---phenylglycine acid chloride, hydrochloride, r-alpha-aminobenzeneacetyl chloride, hydrochloride, r---2-phenylglycine chloride hcl, 2r-amino phenyl acetoyl chloride hydrochloride 1:1
170253
(2R)-2-amino-2-phenylacetyl chloride;hydrochloride
C1=CC=C(C=C1)C(C(=O)Cl)N.Cl

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 177°C
Color Brown
UN Number UN3261
Beilstein 5405183
Formula Weight 206.07
Percent Purity 98%
Quantity 1 g
Flash Point 150°C (302°F)
Sensitivity Moisture sensitive
Physical Form Powder
Chemical Name or Material D-(-)-2-Phenylglycine chloride hydrochloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'