missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Cynomolgus Nectin-4 Protein, His Tag
5341.70 NOK - 39223.36 NOK
Specifications
Accession Number | XP_005541277.1 |
---|---|
Molecular Weight (g/mol) | 36.0 kDa |
Source | Cynomolgus Nectin-4, His Tag (NE4-C52H4) is expressed from human 293 cells (HEK293). It contains AA Gly 32 - Ser 349 (Accession # XP_005541277.1). |
Immunogen | Gly 32 - Ser 349 |
Storage Requirements | -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16437909
|
ACROBiosystems
NE4-C52H4-100UG |
100 μg |
5341.70 NOK
100µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16447909
|
ACROBiosystems
NE4-C52H4-1MG |
1 mg |
39223.36 NOK
1mg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Nectin-4 (gene name PVRL4, poliovirus receptor-like 4) is a 66 kDa type I transmembrane glycoprotein belonging to the Nectin family of Ig superfamily proteins. Nectins are cell adhesion molecules that play a key role in various biological processes such as polarity, proliferation, differentiation and migration, for epithelial, endothelial, immune and neuronal cells, during development and adult life. Nectin-4 is a tumor-associated antigen in 50%, 49% and 86%o of breast, ovarian and lung carcinomas, respectively, mostly on tumors of bad prognosis. Its expression is not detected in the corresponding normal tissues.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 36.0 kDa. The protein migrates as 40-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
XP_005541277.1 | |
Cynomolgus Nectin-4, His Tag (NE4-C52H4) is expressed from human 293 cells (HEK293). It contains AA Gly 32 - Ser 349 (Accession # XP_005541277.1). | |
-20°C | |
Nectin-4 | |
Cynomolgus Monkey | |
PBS, pH7.4 | |
HEK293 | |
Immune Checkpoint Proteins |
36.0 kDa | |
Gly 32 - Ser 349 | |
NECTIN4,LNIR, PRR4, PVRL4 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |