missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropylmagnesium bromide, 0.5M solution in 2-MeTHF, AcroSeal™, Thermo Scientific Chemicals

Catalog Number p-4813442
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
This item is not returnable. View return policy
23719-80-4, 96-47-9
C3H5BrMg
145.28
MFCD00672094
QUPLLJGAJVXXBW-UHFFFAOYSA-M
cyclopropylmagnesium bromide, cyclopropylmagnesiumbromide, bromo cyclopropyl magnesium, magnesium, bromocyclopropyl, cyclopropyl magnesium bromide, cprmgbr, cyclopropyl magnesiumbromide, magnesium,bromocyclopropyl, bromo-cyclopropyl magnesium, cyclopropylmagnesiunn bromide, grignard reagent
11116189
magnesium;cyclopropane;bromide
Br[Mg]C1CC1
This item is not returnable. View return policy
23719-80-4, 96-47-9
C3H5BrMg
145.28
MFCD00672094
QUPLLJGAJVXXBW-UHFFFAOYSA-M
cyclopropylmagnesium bromide, cyclopropylmagnesiumbromide, bromo cyclopropyl magnesium, magnesium, bromocyclopropyl, cyclopropyl magnesium bromide, cprmgbr, cyclopropyl magnesiumbromide, magnesium,bromocyclopropyl, bromo-cyclopropyl magnesium, cyclopropylmagnesiunn bromide, grignard reagent
11116189
magnesium;cyclopropane;bromide
Br[Mg]C1CC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Flash Point −11°C
Packaging AcroSeal™ Glass bottle
Linear Formula C3H5MgBr
Quantity 100 mL
Solubility Information Solubility in water: reacts
Formula Weight 145.28
Chemical Name or Material Cyclopropylmagnesium bromide
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyclopropylmagnesium bromide, 0.5M solution in 2-MeTHF
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H225-Highly flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P223-Do not allow contact with water.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'