Learn More
Cyclopropyl isothiocyanate, 97%, Thermo Scientific Chemicals
Catalog Number p-7045787
Click to view available options
Quantity:
1 g
5 g
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
56601-42-4
C4H5NS
99.15
MFCD00040874
JGFBQFKZKSSODQ-UHFFFAOYSA-N
cyclopropyl isothiocyanate, cyclopropane, isothiocyanato, isothiocyanato-cyclopropane, cyclopropylisothiocyanate, acmc-20albt, pubchem22785, cyclopropyl iso-thiocyanate, cyclopropane,isothiocyanato
92463
isothiocyanatocyclopropane
S=C=NC1CC1
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
56601-42-4
C4H5NS
99.15
MFCD00040874
JGFBQFKZKSSODQ-UHFFFAOYSA-N
cyclopropyl isothiocyanate, cyclopropane, isothiocyanato, isothiocyanato-cyclopropane, cyclopropylisothiocyanate, acmc-20albt, pubchem22785, cyclopropyl iso-thiocyanate, cyclopropane,isothiocyanato
92463
isothiocyanatocyclopropane
S=C=NC1CC1
Cyclopropyl isothiocyanate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Cyclopropyl isothiocyanate is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff fields.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to moisture.
TRUSTED_SUSTAINABILITY
Density 1.07
Boiling Point 87°C to 88°C (35 mmHg)
Flash Point 57°C (134°F)
Refractive Index 1.548
Quantity 1 g
UN Number UN2929
Beilstein 3600974
Sensitivity Moisture sensitive
Formula Weight 99.16
Percent Purity 97%
Chemical Name or Material Cyclopropyl isothiocyanate