Learn More
Cyclopropanesulfonamide, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
Cyclopropanesulfonamide is used as fine chemicals to the pharmaceutical, agrochemical, aroma, polymer and electronic industries.
Solubility
Soluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Cyclopropanesulfonamide is used as fine chemicals to the pharmaceutical, agrochemical, aroma, polymer and electronic industries.
Solubility
Soluble in water.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
Melting Point | 101°C to 106°C |
Quantity | 250 mg |
Solubility Information | Soluble in water. |
Formula Weight | 121.15 |
Percent Purity | 97% |
Chemical Name or Material | Cyclopropanesulfonamide |