Learn More
Cyclopentyl isothiocyanate, 98%, Thermo Scientific™
Catalog Number p-7081673
Click to view available options
Quantity:
1 g
5 g
25 g
33522-03-1
C6H9NS
127.205
MFCD00040877
PJOODZCPFADLCI-UHFFFAOYSA-N
cyclopentyl isothiocyanate, cyclopentane, isothiocyanato, isothiocyanato-cyclopentane, cyclopentanisothiocyanate, cyclopentylisothiocyanate, acmc-20albz
141792
isothiocyanatocyclopentane
C1CCC(C1)N=C=S
33522-03-1
C6H9NS
127.205
MFCD00040877
PJOODZCPFADLCI-UHFFFAOYSA-N
cyclopentyl isothiocyanate, cyclopentane, isothiocyanato, isothiocyanato-cyclopentane, cyclopentanisothiocyanate, cyclopentylisothiocyanate, acmc-20albz
141792
isothiocyanatocyclopentane
C1CCC(C1)N=C=S
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It employed as a intermediate for pharmaceutical.
Solubility
Soluble in ethanol, acetone and hydrolyzes with water.
Notes
Moisture Sensitive, store away form water/moisture. Store in cool, dry conditions in well sealed container. Store away from oxidizing agent. Store under dry inert gas.
TRUSTED_SUSTAINABILITY
Density 1.03
Boiling Point 75°C to 76°C (10 mmHg)
Refractive Index 1.523
Quantity 1 g
UN Number UN2922
Beilstein 2715701
Sensitivity Moisture sensitive
Solubility Information Soluble in ethanol,acetone and hydrolyzes with water.
Formula Weight 127.21
Percent Purity 98%
Chemical Name or Material Cyclopentyl isothiocyanate