missing translation for 'onlineSavingsMsg'
Learn More

Cyclododecyl isothiocyanate, 98%, Thermo Scientific™

Catalog Number p-7058339
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
59037-64-8
C13H23NS
225.394
MFCD00041029
VSCLPFIUOOBLQJ-UHFFFAOYSA-N
cyclododecyl isothiocyanate, acmc-1atsj, cyclododecane,isothiocyanato, cyclododecylisothiocyanate
143589
isothiocyanatocyclododecane
C1CCCCCC(CCCCC1)N=C=S
This item is not returnable. View return policy
59037-64-8
C13H23NS
225.394
MFCD00041029
VSCLPFIUOOBLQJ-UHFFFAOYSA-N
cyclododecyl isothiocyanate, acmc-1atsj, cyclododecane,isothiocyanato, cyclododecylisothiocyanate
143589
isothiocyanatocyclododecane
C1CCCCCC(CCCCC1)N=C=S

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Density 0.99
Refractive Index 1.536
Quantity 1 g
UN Number UN1760
Beilstein 4733327
Sensitivity Moisture sensitive
Formula Weight 225.4
Percent Purity 98%
Chemical Name or Material Cyclododecyl isothiocyanate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'