Learn More
Jackson Immunoresearch™ Cy3 AffiniPure Donkey Anti-Rabbit IgG (H+L)
Brand: Jackson Immunoresearch™ 711-165-152
Additional Details : Weight : 0.01000kg
Specifications
Polyclonal | |
1:100-1:800 | |
Donkey | |
Primary | |
–70°C | |
Whole IgG |
Polyclonal | |
0.01M Sodium Phosphate, 0.25M NaCl, pH 7.6, Stabilizer: 15mg/mL Bovine Serum Albumin (IgG-Free, Protease-Free), 0.05% Sodium Azide Preservative | |
160kDa | |
Bovine, Chicken, Equine, Goat, Guinea Pig, Human, Rat, Sheep, Syrian Hamster, Mouse | |
Cy3 AffiniPure Donkey Anti-Rabbit IgG (H+L) |