missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXCR5 Antibody (614641) [Janelia Fluor™ 669], Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB6198JF669
4607.00 NOK valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
CXCR5 Monoclonal antibody specifically detects CXCR5 in Mouse samples. It is validated for Flow Cytometry,Immunocytochemistry,CyTOF-reported
Specifications
| CXCR5 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| BLR1MDR-15, Burkitt lymphoma receptor 1, Burkitt lymphoma receptor 1, GTP binding protein (chemokine (C-X-C motif)receptor 5), Burkitt lymphoma receptor 1, GTP-binding protein, CD185, CD185 antigen, chemokine (C-X-C motif) receptor 5, C-X-C chemokine receptor type 5, CXC-R5, MDR15CXCR-5, MGC117347, Monocyte-derived receptor 15 | |
| E. coli-derived recombinant mouse CXCR5, Met1-Pro57, Accession # Q04683 | |
| RUO | |
| Primary | |
| Mouse | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, CyTOF | |
| 614641 | |
| 50mM Sodium Borate | |
| Rat | |
| 0.1 mL | |
| Adaptive Immunity, Chemokines and Cytokines, Cytokine Research, GPCR, Immunology, Innate Immunity | |
| 643 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction