missing translation for 'onlineSavingsMsg'
Learn More

CTDSPL2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

1611.84 NOK - 4593.76 NOK

Specifications

Antigen CTDSPL2
Concentration 0.13 mg/mL
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16897434
View Documents Promotion Details
Proteintech
17532-1-AP-20UL
20 μL
1611.84 NOK
20µL
View alternative products
Save Up to 
»
Estimated Shipment: 02-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
16887434
View Documents Promotion Details
Proteintech
17532-1-AP-150UL
150 μL
4593.76 NOK
150µL
View alternative products
Save Up to 
»
Estimated Shipment: 02-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This antibody is a rabbit polyclonal antibody raised against a region of human CTDSPL2.

Probable phosphatase.
Specifications

Specifications

CTDSPL2
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
Rabbit
Human, Mouse, Rat
Q05D32, Q5XIK8, Q8BG15
311368, 329506, 51496
CTDSPL2 Fusion Protein Ag11581
Primary
-20°C
CTDSPL2
0.13 mg/mL
Polyclonal
Liquid
RUO
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
CTDSP like 2, CTDSPL2, HSPC058, HSPC129
Ctdspl2
IgG
Antigen Affinity Chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications