Learn More
Invitrogen™ CSK Recombinant Human Protein
CSK is a tyrosine kinase that has been shown to downregulate the activity of SRC oncoprotein.
Brand: Invitrogen™ PR3080B
Additional Details : Weight : 0.01000kg
Description
CSK downregulates the activity of SRC oncoprotein through phosphorylation of its carboxy terminus. Since cell transformation by SRC is caused by various mechanisms that interfere with this phosphorylation, the CSK gene might function as an antioncogene.
Specifications
NP_004374 | |
50mM Tris, pH 7.5, 100mM NaCl, 10% glycerol | |
100μg | |
RUO - research use only | |
CSK | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
51.9kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
CSK Kinase Family | |
Recombinant | |
E. coli |