Learn More
CPC™ HFC39 Series Quick-Disconnect Coupling Bodies and Inserts
Brand:  CPC™ HFCD22439M
Additional Details : Weight : 0.01360kg
 View more versions of this product
Product Code. 15447173
  • 2934.75 NOK / Pack of 10
Estimated Shipment
27-01-2021
Specifications
USP Class VI, ADCF
125 psi
6.4 mm
2.02 in.
Tubing Connector
-40°F
25.4 mm
Amber
Autoclavable
Polysulfone
51.3 mm
6.4mm
-40°C
Shut-off valves