Learn More
Copper Sulphate solution EU Pharmacopoeia, Fisher Chemical™
Catalog Number 12910854
Click to view available options
Quantity:
1000 mL
Packaging:
Bottle
Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.
Concentration 0.02M
Packaging Bottle
Quantity 1000 mL
Grade Eu. Ph. Reference
Chemical Name or Material Copper Sulphate solution
Product Identifier
  • Copper Sulphate 0.02M solution. European Pharmacopoeia Ref: 3001200 (VL 013)
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P501-Dispose of contents/container to .