Learn More
Copper slug, 6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length, Puratronic™, 99.9999% (metals basis), Alfa Aesar™
Brand: Alfa Aesar 042961.14
Additional Details : CAS Number : 7440-50-8 Weight : 0.02500kg
Quantity | 25g |
---|
Chemical Identifiers
7440-50-8 | |
63.546 | |
RYGMFSIKBFXOCR-UHFFFAOYSA-N | |
23978 | |
copper |
Cu | |
MFCD00010965 | |
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer | |
CHEBI:30052 | |
[Cu] |
Specifications
Copper slug | |
25g | |
7440-50-8 | |
(metals basis) | |
MFCD00010965 | |
RYGMFSIKBFXOCR-UHFFFAOYSA-N | |
copper | |
63.546 | |
CHEBI:30052 | |
99.9999% |
≈3.6g/slug | |
6.35mm (0.25 in.) dia. x 12.7mm (0.50 in.) length | |
99.9999% (metals basis) | |
Cu | |
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer | |
[Cu] | |
Odorless | |
23978 | |
≈3.6g/slug | |
Puratronic™ |