missing translation for 'onlineSavingsMsg'
Learn More

Copper(II) cyclohexanebutyrate, AAS, Cu 15.8%, Thermo Scientific Chemicals

Catalog Number p-7080361
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
2218-80-6
C20H36CuO4
404.05
MFCD00036399
ZGMZTUJLZVTXNR-UHFFFAOYSA-N
cyclohexanebutyric acid, copper salt
131876184
copper;4-cyclohexylbutanoic acid
C1CCC(CC1)CCCC(=O)O.C1CCC(CC1)CCCC(=O)O.[Cu]
This item is not returnable. View return policy
2218-80-6
C20H36CuO4
404.05
MFCD00036399
ZGMZTUJLZVTXNR-UHFFFAOYSA-N
cyclohexanebutyric acid, copper salt
131876184
copper;4-cyclohexylbutanoic acid
C1CCC(CC1)CCCC(=O)O.C1CCC(CC1)CCCC(=O)O.[Cu]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 126°C (decomposition)
Quantity 1 g
Formula Weight 402.01
Grade AAS
Physical Form Powder
Chemical Name or Material Copper (II) cyclohexanebutyrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'