missing translation for 'onlineSavingsMsg'
Learn More

Copper germanide, 99.99% (metals basis), Thermo Scientific™

Catalog Number p-7059912
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
CuGe
MFCD12546178
This item is not returnable. View return policy
CuGe
MFCD12546178

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Physical Form Pieces
Quantity 10 g
Formula Weight 136.16
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Copper germanide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'