missing translation for 'onlineSavingsMsg'
Learn More

Copper flake, -325 mesh, 99% (metals basis), Thermo Scientific Chemicals

Catalog Number p-4408711
Click to view available options
Quantity:
250 g
500 g
1 kg
This item is not returnable. View return policy
7440-50-8
Cu
63.55
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
23978
CHEBI:30052
copper
[Cu]
This item is not returnable. View return policy
7440-50-8
Cu
63.55
MFCD00010965
RYGMFSIKBFXOCR-UHFFFAOYSA-N
cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
23978
CHEBI:30052
copper
[Cu]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 1083°C
Boiling Point 2562°C
Appearance Copper colored flakes
Density 8.94
Assay Percent Range 99% (Metals basis)
Quantity 250 g
Formula Weight 63.55
Mesh Size -325 Mesh
Odor Odorless
Vapor Pressure 0 hPa
Form Flake
Chemical Name or Material Copper
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'